Anti-CD34 Armenian Hamster Monoclonal Antibody (Biotin) [clone: HM34]

Supplier: BioLegend
128603 128604 128603 128604
128603-BLEA 99.68 USD
CA128603-BL CA128604-BL 128603-BL 128604-BL
Anti-CD34 Armenian Hamster Monoclonal Antibody (Biotin) [clone: HM34]
Antibodies
Biotin anti-mouse CD34 [HM34]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: HM34
Conjugation: Biotin
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR