Anti-CDK2AP2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP19378C
89514-858EA 636.79 USD
89514-858
Anti-CDK2AP2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CDK2AP2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR