Anti-CD197 (CCR7) Rat Monoclonal Antibody (Biotin) [clone: 4B12]

Supplier: BioLegend
120104 120104 120103 120103
120104-BLEA 443.52 USD
120104-BL CA120104-BL 120103-BL CA120103-BL
Anti-CD197 (CCR7) Rat Monoclonal Antibody (Biotin) [clone: 4B12]
Antibodies
Biotin anti-mouse CD197 (CCR7) [4B12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD197 (CCR7)
Clonality: Monoclonal
Clone: 4B12
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR