Anti-CD16/32 Rat Monoclonal Antibody (Biotin) [clone: 93]

Supplier: BioLegend
101303 101303
101303-BLEA 89.58 USD
CA101303-BL 101303-BL
Anti-CD16/32 Rat Monoclonal Antibody (Biotin) [clone: 93]
Antibodies
Biotin anti-mouse CD16/32 [93]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD16/32
Clonality: Monoclonal
Clone: 93
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR