Anti-IgG Isotype Ctrl Syrian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: SHG-1]

Supplier: BioLegend
402012 402012
402012-BLEA 229.93 USD
402012-BL CA402012-BL
Anti-IgG Isotype Ctrl Syrian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: SHG-1]
Antibodies
APC Syrian Hamster IgG Isotype Ctrl [SHG-1]; Isotype: Syrian Hamster IgG; Reactivity: Unknown; Apps: FC, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG Isotype Ctrl
Clonality: Monoclonal
Clone: SHG-1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Syrian Hamster
Isotype: Syrian Hamster IgG
Reactivity: Unknown
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR