Anti-CTNND1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP20722B
10218-418EA 580.69 USD
10218-418
Anti-CTNND1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CTNND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR