Anti-TCR a/b Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: R73]

Supplier: BioLegend
201110 201110
201110-BLEA 332.48 USD
CA201110-BL 201110-BL
Anti-TCR a/b Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: R73]
Antibodies
APC anti-rat TCR αβ [R73]; Isotype: Mouse (BALB/c) IgG1, κ; Reactivity: Rat, Cross-Reactivity: Cynomolgus, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR a/b
Clonality: Monoclonal
Clone: R73
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG1, κ
Reactivity: Rat;Cross-Reactivity: Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR