Anti-CD11b/c Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OX-42]

Supplier: BioLegend
201809 201809
201809-BLEA 192.08 USD
201809-BL CA201809-BL
Anti-CD11b/c Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OX-42]
Antibodies
APC anti-rat CD11b/c [OX-42]; Isotype: Mouse IgG2a, κ; Reactivity: Rat; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11b/c
Clonality: Monoclonal
Clone: OX-42
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR