Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: C398.4A]

Supplier: BioLegend
313509 313510 313509 313510
313509-BLEA 128.45 USD
313509-BL CA313509-BL CA313510-BL 313510-BL
Anti-CD278 (ICOS) Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: C398.4A]
Antibodies
APC anti-human/mouse/rat CD278 (ICOS) [C398.4A]; Isotype: Armenian Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Mouse, Rat, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that reagents be titrated for optimal performance in the particular application.

Type: Primary
Antigen: CD278 (ICOS)
Clonality: Monoclonal
Clone: C398.4A
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Human;African Green;Baboon;Cynomolgus;Mouse;Rat;Rhesus;Swine (Pig;Porcine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR