Anti-IgM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MHM-88]

Supplier: BioLegend
314509 314510 314509 314510
314510-BLEA 314.1 USD
314510-BL 314509-BL CA314509-BL CA314510-BL
Anti-IgM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MHM-88]
Antibodies
APC anti-human IgM [MHM-88]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. <strong>Test size products are transitioning from 20 μl to 5 μl per test</strong>. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume. It is recommended that the reagent be titrated for optimal performance for each application. Read more at <a href="/testsize" target="_blank">www.biolegend.com/testsize</a> regarding the test size change.

Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone: MHM-88
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR