Anti-CBFA2T2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP16252A
89520-100EA 637.11 USD
89520-100
Anti-CBFA2T2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:2000, Immunofluorescence: 1:10~50,

Type: Primary
Antigen: CBFA2T2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR