Anti-MAP3K13 Mouse Monoclonal Antibody [clone: 810CT26.6.1.1]

Supplier: Abcepta
AM2234B
10217-778EA 552.16 USD
10217-778
Anti-MAP3K13 Mouse Monoclonal Antibody [clone: 810CT26.6.1.1]
Antibodies
Western Blot: 1:1000, Immunofluorescence: 1:100,

Type: Primary
Antigen: MAP3K13
Clonality: Monoclonal
Clone: 810CT26.6.1.1
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR