Anti-CD318 (CDCP1) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CUB1]

Supplier: BioLegend
324008 324008
324008-BLEA 339 USD
324008-BL CA324008-BL
Anti-CD318 (CDCP1) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CUB1]
Antibodies
APC anti-human CD318 (CDCP1) [CUB1]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD318 (CDCP1)
Clonality: Monoclonal
Clone: CUB1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR