Anti-CD274 (B7-H1, PD-L1) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 29E.2A3]

Supplier: BioLegend
329708 329707 329707 329708
329708-BLEA 415.24 USD
329708-BL 329707-BL CA329707-BL CA329708-BL
Anti-CD274 (B7-H1, PD-L1) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 29E.2A3]
Antibodies
APC anti-human CD274 (B7-H1, PD-L1) [29E.2A3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. <strong>Test size products are transitioning from 20 μl to 5 μl per test</strong>. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at <a href="/testsize" target="_blank">www.biolegend.com/testsize</a> regarding the test size change.

Type: Primary
Antigen: CD274 (B7-H1, PD-L1)
Clonality: Monoclonal
Clone: 29E.2A3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR