Anti-PDPK1 Mouse Monoclonal Antibody [clone: 340CT15.1.1]

Supplier: Abcepta
AM1954B
89521-020EA 637.11 USD
89521-020
Anti-PDPK1 Mouse Monoclonal Antibody [clone: 340CT15.1.1]
Antibodies
Western Blot: 1:100~250,

Type: Primary
Antigen: PDPK1
Clonality: Monoclonal
Clone: 340CT15.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR