Anti-TLR3 Mouse Monoclonal Antibody [clone: 303CT19.9.2]

Supplier: Abcepta
AM1944B
89521-006EA 603.19 USD
89521-006
Anti-TLR3 Mouse Monoclonal Antibody [clone: 303CT19.9.2]
Antibodies
Western Blot: 1:500,

Type: Primary
Antigen: TLR3
Clonality: Monoclonal
Clone: 303CT19.9.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR