Anti-BIRC5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-A34 200-401-A34
RL200-401-A34EA 518.29 USD
CA200-401-A34 RL200-401-A34
Anti-BIRC5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR