Anti-SEPHS1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-A22 100-401-A22
RL100-401-A22EA 518.16 USD
CA100-401-A22 RL100-401-A22
Anti-SEPHS1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SEPHS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR