Anti-PMS2 Mouse Monoclonal Antibody [clone: 1204]

Supplier: Rockland Immunochemicals
200-301-864 200-301-864
RL200-301-864EA 528.35 USD
RL200-301-864 CA200-301-864
Anti-PMS2 Mouse Monoclonal Antibody [clone: 1204]
Antibodies
Type: Primary
Antigen: PMS2
Clonality: Monoclonal
Clone: 1204
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Hamster, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR