Anti-NSP9 Mouse Monoclonal Antibody [clone: 2C6.H1]

Supplier: Rockland Immunochemicals
200-301-A56 200-301-A56
RL200-301-A56EA 657.14 USD
RL200-301-A56 CA200-301-A56
Anti-NSP9 Mouse Monoclonal Antibody [clone: 2C6.H1]
Antibodies
Type: Primary
Antigen: NSP9
Clonality: Monoclonal
Clone: 2C6.H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Hepatitis Virus, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR