Anti-CFLAR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-983 200-401-983
RL200-401-983EA 518.29 USD
CA200-401-983 RL200-401-983
Anti-CFLAR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CFLAR
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR