Anti-CYTIP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-A23 100-401-A23
RL100-401-A23EA 518.16 USD
RL100-401-A23 CA100-401-A23
Anti-CYTIP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYTIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR