Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Mouse Polyclonal Antibody [clone: 17F6.B11]

Supplier: Rockland Immunochemical
200-301-268S
Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone: 17F6.B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 µL 200-301-268S RL200-301-268S Each (25ml) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol AKT1
Antigen Name V-Akt Murine Thymoma Viral Oncogene Homolog 1
Reactivity Rat, Human, Mouse, Monkey
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Clone 17F6.B11
Host Mouse
Isotype IgG1 kappa
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Antigen Synonyms PKB beta, PKB-GAMMA, RAC-beta serine/threonine-protein kinase, RAC, RAC-gamma serine/threonine-protein kinase, PKBB, PRKBG, Murine thymoma viral (v-akt) homolog-2, RAC-PK-gamma, AKT2, STK-2, PKBG, rac protein kinase beta, kinase Akt1, EC 2.7.11.1, RAC-gamma serine/threonine protein kinase, RAC-PK-alpha, RAC-alpha serine/threonine kinase, PRKBB, PKB-alpha, Protein kinase B, v-akt murine thymoma viral oncogene homolo, Protein kinase B beta, RAC-gamma, Protein kinase Akt-2, PKB gamma, AKT1 kinase, RAC-PK-beta, C-AKT, PKBBETA, PKB, RAC-BETA, AKT
Gene ID 207
Modification phospho-Ser473
Concentration 1.0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross Adsorption No
Immunogen Anti-AKT pS473 antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding S473 of human AKT1 Protein, followed by hybridoma development
Purification Protein A purified
Form Liquid (sterile filtered)
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
Storage Temperature -20°C
Shipping Temperature Dry Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty