Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Mouse Polyclonal Antibody [clone: 14E5.16C8.25F6]

Supplier: Rockland Immunochemical
200-301-401
Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone: 14E5.16C8.25F6
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype:
Reactivity: Chimpanzee, Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 200-301-401 RL200-301-401 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol AKT1
Antigen Name V-Akt Murine Thymoma Viral Oncogene Homolog 1
Reactivity Rat, Human, Chimpanzee, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Clone 14E5.16C8.25F6
Host Mouse
Isotype IgG1 kappa
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow Cytometry Yes
Antigen Synonyms PKB, EC 2.7.11.1, AKT1 kinase, PKB gamma, PKB-GAMMA, RAC-gamma, PKB-alpha, AKT2, RAC-alpha serine/threonine kinase, AKT, v-akt murine thymoma viral oncogene homolo, RAC-gamma serine/threonine-protein kinase, Protein kinase B beta, PKB beta, PKBBETA, C-AKT, RAC-PK-alpha, RAC-PK-gamma, PRKBB, RAC, RAC-gamma serine/threonine protein kinase, PKBB, kinase Akt1, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, STK-2, PKBG, PRKBG, rac protein kinase beta, Murine thymoma viral (v-akt) homolog-2, Protein kinase Akt-2, Protein kinase B
Gene ID 207
Epitope Center
Concentration 1.0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross Adsorption No
Immunogen Anti-AKT Antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues internal to the human AKT1 Protein.
Purification Protein A purified
Form Liquid (sterile filtered)
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
Storage Temperature -20°C
Shipping Temperature Dry Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty