Anti-AcK Mouse Polyclonal Antibody [clone: 7F8]

Supplier: Rockland Immunochemicals
200-301-A97 200-301-A97
RL200-301-A97EA 528.25 USD
CA200-301-A97 RL200-301-A97
Anti-AcK Mouse Polyclonal Antibody [clone: 7F8]
Antibodies
Type: Primary
Antigen: AcK
Clonality: Polyclonal
Clone: 7F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mammals, Avian
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR