Anti-EIF2AK3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-962 100-401-962
RL100-401-962EA 522.84 USD
RL100-401-962 CA100-401-962
Anti-EIF2AK3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EIF2AK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR