Anti-IgG1, k Isotype Ctrl Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: RTK2071]

Supplier: BioLegend
400420 400420
400420-BLEA 193.1 USD
400420-BL CA400420-BL
Anti-IgG1, k Isotype Ctrl Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: RTK2071]
Antibodies
Alexa Fluor® 700 Rat IgG1, κ Isotype Ctrl [RTK2071]; Isotype: Rat IgG1, κ; Reactivity: Rat; Apps: FC, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. Use our <a href="/concentrationlookup" target="_blank">Concentration Lookup</a> tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2071
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR