Anti-CD90.2 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 30-H12]

Supplier: BioLegend
105319 105319 105320 105320
105319-BLEA 109.03 USD
105319-BL CA105319-BL CA105320-BL 105320-BL
Anti-CD90.2 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 30-H12]
Antibodies
Alexa Fluor® 700 anti-mouse CD90.2 [30-H12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Does not react with Thy-1.1 (CD90.1); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse;Does not react with Thy-1.1 (CD90.1)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR