Anti-CD86 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: PO3]

Supplier: BioLegend
105122 105122
105122-BLEA 247.16 USD
105122-BL CA105122-BL
Anti-CD86 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: PO3]
Antibodies
Alexa Fluor® 700 anti-mouse CD86 [PO3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: PO3
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR