Anti-CD45R Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: RA3-6B2]

Supplier: BioLegend
103232 103231 103231 103232
103231-BLEA 105.39 USD
CA103232-BL CA103231-BL 103231-BL 103232-BL
Anti-CD45R Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: RA3-6B2]
Antibodies
Alexa Fluor® 700 anti-mouse/human CD45R/B220 [RA3-6B2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cat (Feline); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Human;Cross-Reactivity: Cat (Feline)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR