Anti-CD45.1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: A20]

Supplier: BioLegend
110724 110723 110724 110723
110724-BLEA 321.15 USD
CA110723-BL CA110724-BL 110724-BL 110723-BL
Anti-CD45.1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: A20]
Antibodies
Alexa Fluor® 700 anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: Mouse (A.SW) IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR