Anti-RRN3P2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP19182B
89515-242EA 636.79 USD
89515-242
Anti-RRN3P2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: RRN3P2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR