Anti-CD197 (CCR7) Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 4B12]

Supplier: BioLegend
120112 120110 120112 120110
120112-BLEA 158.69 USD
CA120112-BL 120112-BL 120110-BL CA120110-BL
Anti-CD197 (CCR7) Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 4B12]
Antibodies
Alexa Fluor® 488 anti-mouse CD197 (CCR7) [4B12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106cells in 100 μl staining volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD197 (CCR7)
Clonality: Monoclonal
Clone: 4B12
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR