Anti-TCR alpha/beta Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: IP26]

Supplier: BioLegend
306711 306712 306711 306712
306711-BLEA 133.83 USD
CA306711-BL CA306712-BL 306711-BL 306712-BL
Anti-TCR alpha/beta Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: IP26]
Antibodies
Alexa Fluor® 488 anti-human TCR α/β [IP26]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR alpha/beta
Clonality: Monoclonal
Clone: IP26
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR