Anti-HLA-DR Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: LN3]

Supplier: BioLegend
327010 327010
327010-BLEA 285.98 USD
327010-BL CA327010-BL
Anti-HLA-DR Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: LN3]
Antibodies
Alexa Fluor® 488 anti-human HLA-DR [LN3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: LN3
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR