Anti-CD51/61 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 23C6]

Supplier: BioLegend
304408 304408
304408-BLEA 285.98 USD
304408-BL CA304408-BL
Anti-CD51/61 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 23C6]
Antibodies
Alexa Fluor® 488 anti-human CD51/61 [23C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Chicken, Cynomolgus, Rabbit (Lapine); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD51/61
Clonality: Monoclonal
Clone: 23C6
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Chicken;Cynomolgus;Rabbit (Lapine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR