Anti-CD340 (erbB2/HER-2) Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 24D2]

Supplier: BioLegend
324410 324410
324410-BLEA 394.78 USD
324410-BL CA324410-BL
Anti-CD340 (erbB2/HER-2) Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 24D2]
Antibodies
Alexa Fluor® 488 anti-human CD340 (erbB2/HER-2) [24D2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IF; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD340 (erbB2/HER-2)
Clonality: Monoclonal
Clone: 24D2
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR