Anti-ZIC2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-848 600-401-848
RL600-401-848EA 518.16 USD
RL600-401-848 CA600-401-848
Anti-ZIC2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR