Anti-TLR9 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-A69 600-401-A69
RL600-401-A69EA 518.29 USD
CA600-401-A69 RL600-401-A69
Anti-TLR9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TLR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR