Anti-THRA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-A38S 600-401-A38S
RL600-401-A38SEA 147.5 USD
RL600-401-A38S CA600-401-A38S
Anti-THRA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: THRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR