Anti-SLIT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-860 600-401-860
RL600-401-860EA 522.84 USD
CA600-401-860 RL600-401-860
Anti-SLIT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLIT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR