Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PARP p85 Fragment Rabbit Polyclonal Antibody

Supplier: Promega Corporation
G7341
Specific in situ marker of apoptosis recognizing the p85 fragment of PARP resulting from caspase-3 cleavage.

  • A Specific In Situ Marker of Apoptosis
  • Specifically detects PARP p85 fragment in human, rat and bovine cells and tissues

Poly (ADP-ribose) polymerase (PARP), a nuclear enzyme involved in DNA repair, is a well known substrate for caspase-3 cleavage during apoptosis. Anti-PARP p85 Fragment pAb is a rabbit polyclonal antibody specific for the p85 fragment of PARP that results from caspase cleavage of the 116kDa intact molecule and thus provides an in situ marker for apoptosis. The antibody is affinity-purified using a peptide that corresponds to a region of the p85 fragment of PARP. The PARP immunogen is a synthetic peptide, gly-val-asp-glu-val-ala-lys (GVDEVAK), representing the N-terminus of the large C-terminal fragment of human PARP that results from caspase-3 cleavage. Each batch of antibody is quality assurance tested for use in immunostaining applications and contains sufficient antibody for 50 immunocytochemical reactions at the suggested working dilution of 1:100.

It is quality assurance tested for use in immunostaining applications and contains sufficient antibody for 50 immunocytochemical reactions at the suggested working dilution of 1:100.

Type:
Antigen: PARP p85 Fragment
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: C-terminal, N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Bovine cells and Tissues




 

ORDER

Expand All / Collapse All
Size Storage Temperature Supplier No. VWR Catalog Number Unit Price Quantity
50 μl -30°C to -10°C G7341 PAG7341 Each (50µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PARP1
Antigen Name Poly (ADP-ribose) polymerase 1
Reactivity Bovine, Human, Rat
Clonality Polyclonal
Host Rabbit
Western Blot Yes
Antigen Synonyms pADPRT-1, ADPRT, PARP1, PARP-1, ADPRT1, PARP, PPOL
Epitope C-terminal, N-terminal
Molecular Weight 116 kDa
Tested Applications Immunohistochemistry

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty