Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: 53-6.7]

Supplier: ProSci
76-690
10765-578EA 448.21 USD
10765-578
Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy5®) [clone: 53-6.7]
Antibodies
Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE-Cy5
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR