Anti-LAMP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: H4A3]

Supplier: ProSci
76-893
10764-982EA 683.4 USD
10764-982
Anti-LAMP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: H4A3]
Antibodies
The H4A3 monoclonal antibody specifically reacts with human CD107a, a heavily glycosylated type I membrane glycoprotein. CD107a is also known as Lysosomal-associated membrane protein 1 (LAMP-1) and is widely expressed intracellular antigen. It can be found on the surface of PHA-activated lymphocytes, activated platelets, cytotoxic T cells, NK cells, macrophages, epithelial cells, endothelial cells, and some tumor lines. CD107a is a ligand for E-selectin and galaptin and is reported to be involved in cell adhesion and tumor metastasis.

Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: H4A3
Conjugation: APC
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR