Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGAX Hamster Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5®.5) [clone: N418]

Supplier: Prosci
The N418 monoclonal antibody specifically reacts with the integrin alpha x chain of the mouse CD11c, which is expressed on dendritic cells, CD4-/CD8+ intestinal intraepithelial lymphocytes (IEL) and some activated T lymphocytes. Low levels of CD11c were detected on mouse splenic natural killer cells and on the monocyte/macrophage lineage cells.CD11c expression is upregulated on IEL and T lymphocytes after activation. It binds to CD54, fibrinogen and iC3b and influences the leukocyte adhesive interactions. The N418 antibody binds to CD11c on splenic dendritic cells of the mouse in the T-dependent areas. It also contributes to the binding of iC3b.

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: N418
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5®.5
Host: Hamster
Isotype: IgG
Reactivity: Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 76-107 10766-218 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol ITGAX
Antigen Name Integrin Alpha 10
Reactivity Mouse
Conjugation PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Clonality Monoclonal
Antibody Type Primary
Clone N418
Host Hamster
Isotype IgG
Flow Cytometry Yes
Antigen Synonyms N418, integrin aX, AI449405, Cd11c, Cr4, ITGAX, integrin alpha-X, CD11 antigen-like family member C
Concentration 0.2 mg/ml
Cross Adsorption No
Purification purified utilizing affinity chromatography and unreacted dye was removed from the product.
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty