Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FCER1A Hamster Monoclonal Antibody [clone: FceR1]

Supplier: Prosci
77-174
The MAR-1 antibody reacts with the Fc epsilon Receptor I alpha chain (FceRIa), a transmembrane protein member of the Ig superfamily. This chain, together with a beta chain and two gamma chains form a tetrameric complex that supports IgE-mediated signaling and subsequent release of chemical mediators of allergy and immediate hypersensitivity. FceR1a is upregulated in the presence of IgE on those cell types which express it, such as Mast cells and Basophils.The MAR-1 antibody is widely used both in flow cytometry and for depletion of cells in vitro / in vivo.

Type: Primary
Antigen: FCER1A
Clonality: Monoclonal
Clone: FceR1
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 77-174 10765-840 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol FCER1A
Antigen Name Fc fragment of IgE receptor Ia
Reactivity Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone FceR1
Host Hamster
Isotype IgG
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow Cytometry Yes
Antigen Synonyms FcERI, FCE1A
Concentration 2 mg/ml
Format purified antibody
Cross Adsorption No
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty