Anti-NOTCH1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: mN1A]

Supplier: ProSci
76-998
10765-898EA 535.13 USD
10765-898
Anti-NOTCH1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: mN1A]
Antibodies
The mN1A monoclonal antibody reacts with the intracellular domain of human and mouse Notch1 with high affinity for the activated intracellular form and lower affinity for the full length heterodimeric Notch1 forms. It does not react with Notch2,3, or 4 and does not cross-react with rat thymocytes. The neurogenic locus homolog protein 1 (Notch1) is a trasmembrane receptor that regulates cell fate decisions. It is expressed in CD4-CD8- and CD4-CD8+ thymocytes, and plays a role in the regulation of myelopoiesis and lymphopoiesis.

Type: Primary
Antigen: NOTCH1
Clonality: Monoclonal
Clone: mN1A
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR