Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2.43]

Supplier: ProSci
76-649
10764-726EA 345.65 USD
10764-726
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2.43]
Antibodies
The 2.43 monoclonal antibody specifically reacts with mouse CD8 antigen. CD8a (the alpha chains) form heterodimers with CD8b (the beta chains) or homodimers (alpha-alpha), which occur as receptors on the surface of the majority of thymocytes. A subpopulation of mature T lymphocytes expresses the CD8 alpha beta (alpha beta TCR T cells), and a subpopulation of intestinal intraepithelial lymphocytes and dendritic cells express CD8a without CD8b. CD8 interacts with the mouse major histocompatibility complex class I (MHC class I) molecules on antigen-presenting cells or epithelial cells.

Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: 2.43
Conjugation: FITC
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR