Anti-CD8A Rat Monoclonal Antibody (violetFluor® 450) [clone: 2.43]

Supplier: ProSci
76-647
10764-722EA 535.12 USD
10764-722
Anti-CD8A Rat Monoclonal Antibody (violetFluor® 450) [clone: 2.43]
Antibodies
The 2.43 monoclonal antibody specifically reacts with mouse CD8 antigen. CD8a (the alpha chains) form heterodimers with CD8b (the beta chains) or homodimers (alpha-alpha), which occur as receptors on the surface of the majority of thymocytes. A subpopulation of mature T lymphocytes expresses the CD8 alpha beta (alpha beta TCR T cells), and a subpopulation of intestinal intraepithelial lymphocytes and dendritic cells express CD8a without CD8b. CD8 interacts with the mouse major histocompatibility complex class I (MHC class I) molecules on antigen-presenting cells or epithelial cells. BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: 2.43
Conjugation: Violet-450
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR