Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UCHT1]

Supplier: Prosci
76-296
The UCHT1 monoclonal antibody specifically reacts with the epsilon chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains gamma, delta, and epsilon chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction. Unlike HIT3a, another specific antibody of CD3, the UCHT1 antibody can stain both the surface and intracellular CD3 epsilon. The immobilized UCHT1 can cross-link with the TCR complex, enhancing cellular activation and proliferation.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: UCHT1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 Tests 76-296 10764-320 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CD3E
Antigen Name CD3 Molecule Epsilon
Reactivity Human
Conjugation APC (Allophycocyanin)
Clonality Monoclonal
Antibody Type Primary
Clone UCHT1
Host Mouse
Isotype IgG1 kappa
Flow Cytometry Yes
Antigen Synonyms T-cell surface antigen T3/Leu-4 epsilon chain, CD3e antigen, epsilon polypeptide TiT3 complex, FLJ18683, T3E, T-cell surface glycoprotein CD3 epsilon chain, epsilon subunit of T3, T-cell antigen receptor complex, TCRE, CD3-epsilon
Concentration 5 uL (0.25 ug) / test
Cross Adsorption No
Purification purified utilizing affinity chromatography and unreacted dye was removed from the product.
Form liquid
Storage Buffer Phosphate-buffered aqueous solution, ?0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
Storage Temperature The product should be stored undiluted at 4°C and should be protected from prolonged exposure to light. Do not freeze.
Shipping Temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty